missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco Labs Pack of 12 Sterile Wide Mouth Amber HDPE Reagent Bottles with Polypropylene Caps
Description
- Amber reagent bottles: Includes 12 sterile wide-mouth HDPE bottles with polypropylene caps, designed for laboratory sample storage and secure transportation.
- UV light protection: Amber color shields light-sensitive samples from harmful UV rays, ideal for protecting reagents, solutions, and light-sensitive substances.
- Leak-proof design: Bottles are rigorously leak-proof tested to ensure safe storage and transport of chemicals, liquids, or reagents in laboratory and industrial settings.
- Chemical-resistant materials: Manufactured from durable HDPE with excellent chemical resistance, suitable for diverse applications in laboratories and industries.
- Easy to handle and transport: Wide-mouth design allows easy filling and pouring.
Specifications
Specifications
| Capacity (Metric) | 250 mL |
| Capacity (English) | 8.45 oz. |
| Color | Amber |
| Diameter (Metric) Inner Neck | 33.1 mm |
| Material | HDPE |
| No. per Pack | 12 Pk. |
| Sterility | Sterile |
| Closure Included | Yes |
| Graduated | No |
| Mouth | Wide |
| Show More |