missing translation for 'onlineSavingsMsg'
Learn More

Eisco Labs Pack of 12 Sterile Wide Mouth Amber HDPE Reagent Bottles with Polypropylene Caps

Catalog No. S000130
Change view
Click to view available options
Capacity (Metric):
30 mL
125 mL
250 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S000130 30 mL
S000131 125 mL
S000132 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S000130 Supplier Eisco Labs Supplier No. CH5501PK12
Only null left

Pack of 12 sterile wide-mouth amber HDPE reagent bottles with polypropylene caps areUV-protective, leak-tested, and chemically resistant, ideal for safe storage and transfer of light-sensitive liquids in lab and industrial settings.

  • Amber reagent bottles: Includes 12 sterile wide-mouth HDPE bottles with polypropylene caps, designed for laboratory sample storage and secure transportation.
  • UV light protection: Amber color shields light-sensitive samples from harmful UV rays, ideal for protecting reagents, solutions, and light-sensitive substances.
  • Leak-proof design: Bottles are rigorously leak-proof tested to ensure safe storage and transport of chemicals, liquids, or reagents in laboratory and industrial settings.
  • Chemical-resistant materials: Manufactured from durable HDPE with excellent chemical resistance, suitable for diverse applications in laboratories and industries.
  • Easy to handle and transport: Wide-mouth design allows easy filling and pouring.

Specifications

Capacity (Metric) 30 mL
Capacity (English) 1 oz.
Color Amber
Diameter (Metric) Inner Neck 21.6 mm
Material HDPE
No. per Pack 12 Pk.
Sterility Sterile
Closure Included Yes
Graduated No
Mouth Wide
Height (Metric) 62 mm
Product Type Reagent Bottle
Diameter (Metric) 34.8 mm
Autoclavable Not Autoclavable
Show More Show Less