missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Pacific Blue™ C5-Maleimide

Catalog No. 501121576
Encompass
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501121576 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501121576 Supplier Molecular Probes™ Supplier No. P30506

Pacific Blue™ C5-maleimide

Pacific Blue™ C5-maleimide is an excellent reagent for thiol-selective modification, quantitation and analysis. In this reaction, the thiol is added across the double bond of the maleimide to yield a thioether. This compound does not react with methionine, histidine or tyrosine. Reaction of Pacific Blue™ C5-maleimide with amines usually requires a higher pH than reaction of maleimides with thiols.

View all Pacific Blue™ dye products..

View the Fluorophore Selection Guide.

Specifications

Format Solid
Quantity 1 mg
Product Type Maleimide
Solubility DMSO (Dimethylsulfoxide)
Content And Storage Contains 1 vial of Pacific Blue™ dye (1 mg). Store in freezer (-5 to -30°C) and protect from light.
Chemical Reactivity Thiol
Detection Method Fluorescence
Flow Cytometer Laser Lines 405
Shipping Condition Room Temperature
Reactive Moiety Maleimide
Label or Dye Pacific Blue™
Label Type Pacific Dyes
Color Off-White
Product Line Molecular Probes, Pacific Blue
Emission 455
Excitation 410
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.