missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Oxymetazoline hydrochloride

CAS: 2315-02-8 | C16H25ClN2O

$239.73 - $239.73

Chemical Identifiers

CAS 2315-02-8
Molecular Formula C16H25ClN2O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC463390050
View Documents
Thermo Scientific Chemicals
463390050
5 g
Each for $239.73
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2315-02-8
C16H25ClN2O

Specifications

Oxymetazoline hydrochloride
C16H25ClN2O
5 g
2315-02-8
296.84
Videos
Safety and Handling

Safety and Handling

GHS H Statement Fatal if swallowed.

Causes serious eye damage.

Harmful to aquatic life with long lasting effects.

GHS P Statement Rinse mouth.

Wear eye protection/face protection.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

Immediately call a POISON CENTER or doctor/physician.

GHS Signal Word: Danger

EINECSNumber : 219-015-0

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only