CAS RN 2315-02-8
IUPAC Name:
hydrogen 6-tert-butyl-3-[(4,5-dihydro-1H-imidazol-2-yl)methyl]-2,4-dimethylphenol chloride
Synonyms:
oxymetazoline hydrochloride
Molecular Weight (g/mol):
296.84
Molecular Formula:
C16H25ClN2O
InChi Key:
BEEDODBODQVSIM-UHFFFAOYSA-N
SMILES:
[H+].[Cl-].CC1=CC(=C(O)C(C)=C1CC1=NCCN1)C(C)(C)C
1
–
1
of
1
results
1
Promotions Available
Thermo Scientific Chemicals Oxymetazoline hydrochloride
Thermo Scientific Chemicals Oxymetazoline hydrochloride
CAS: 2315-02-8 Molecular Formula: C16H25ClN2O