missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Oxymetazoline hydrochloride

Catalog No. AC463390050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC463390050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC463390050 Supplier Thermo Scientific Chemicals Supplier No. 463390050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2315-02-8 | C16H25ClN2O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2315-02-8
Molecular Formula C16H25ClN2O

Specifications

Product Type Oxymetazoline hydrochloride
Formula Weight 296.84
Quantity 5 g
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Fatal if swallowed.

Causes serious eye damage.

Harmful to aquatic life with long lasting effects.

Precautionary Statement GHS P Statement Rinse mouth.

Wear eye protection/face protection.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

Immediately call a POISON CENTER or doctor/physician.

EINECSNumber 219-015-0

RUO – Research Use Only