missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Oxacillin sodium salt, 95%

Catalog No. AC455440010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC455440010 1 g
AC455440050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC455440010 Supplier Thermo Scientific Chemicals Supplier No. 455440010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Oxacillin sodium salt, CAS # 1173-88-2, is an antibiotic that is used primarily in studying gram-positive bacteria.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Oxacillin sodium salt is a narrow-spectrum beta-lactam antibiotic
  • It inhibits biosynthesis of bacterial cell walls at the level of transpeptidation (PBP) of peptidoglycans

Applications

  • Oxacillin sodium salt is an antibiotic that is active against gram-positive bacteria
  • Oxacillin can be used in studies of mechanisms of resistance to penicillinase and antimicrobial susceptibility
  • It has also been used to study autolysins

Chemical Identifiers

CAS 1173-88-2
Molecular Formula C19H21N3NaO5S
Molecular Weight (g/mol) 426.443
InChI Key IFFSBWVMGGQKTD-VICXVTCVSA-N
PubChem CID 131673929
IUPAC Name (2S,5R,6R)-3,3-dimethyl-6-[(5-methyl-3-phenyl-1,2-oxazole-4-carbonyl)amino]-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;molecular hydrogen;sodium
SMILES [HH].CC1=C(C(=NO1)C2=CC=CC=C2)C(=O)NC3C4N(C3=O)C(C(S4)(C)C)C(=O)O.[Na]

Specifications

Type Oxacillin Sodium Salt
Infrared Spectrum Authentic
Water 3.5 to 5.0%
Quantity 1 g
Solubility Information Solubility in water: 50g/L.
Formula Weight 423.42
Percent Purity 95%
Chemical Name or Material Oxacillin sodium salt

RUO – Research Use Only