missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Osmium tetroxide, 4 wt.% solution in water
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
Spécifications
Spécifications
| Boiling Point | 100.0°C |
| Chemical Name or Material | Osmium tetroxide |
| Color | Yellow |
| CAS | 20816-12-0, 7732-18-5 |
| Physical Form | Solution |
| Quantity | 5 mL |
| Molecular Formula | O4Os |
| Linear Formula | OsO4 |
| MDL Number | MFCD00011150 |
| Fieser | 01,759; 02,301; 04,361; 05,141; 06,424; 10,290; 12,358; 13,186; 14,235; 15,240; 16,249; 17,236 |
| Afficher plus de résultats |
RUO – Research Use Only