missing translation for 'onlineSavingsMsg'
Learn More

ORP Standard, +200 mV, Ricca Chemical

Catalog No. p-2780121
Change view
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
546454 120 mL Amber Glass Bottle
5464516 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 546454 Supplier Ricca Chemical Company Supplier No. 5464.54
Only null left

4 M KCl saturated with AgCl filling solution

Chemical Identifiers

CAS 14459-95-1 , 13746-66-2 , 7447-40-7 , 7732-18-5
Molecular Formula C6H6FeK4N6O3
Molecular Weight (g/mol) 422.39
MDL Number MFCD00167023
InChI Key NLYKFUFIUWPPIZ-UHFFFAOYSA-N
IUPAC Name tetrapotassium hexacyanoironbis(ylium) trihydrate
SMILES O.O.O.[K+].[K+].[K+].[K+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)C#N

Specifications

Packaging Amber Glass Bottle
Concentration or Composition Notes 4M KCl saturated with AgCl filling solution
CAS Min % 97.0, 0.73, 0.29, 0.03
CAS Max % 100.0, 0.76, 0.3, 0.03
Boiling Point 100°C
Quantity 120 mL
Chemical Name or Material Oxidation-Reduction Potential Standard, +200mV vs. Ag/AgCl