missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ Combination pH Electrodes

Environmentally friendly lab pH electrodes to meet RoHS requirements by containing no lead, mercury or other hazardous substances — making disposal of old electrodes easy and hassle free.

$636.73 - $1,004.39

Spécifications

Material Epoxy Body
pH Range 0 to 14
Temperature (Metric) Operating 0°C to 90°C
For Use With (Equipment) Orion pH Meter
Length (English) Cable 39.3 in.
voir les spécifications

Produits 3
Numéro de catalogue Numéro du manufacturier. Filling Solution Type Junction Prix Quantity & Availability  
Numéro de catalogue Numéro du manufacturier. Filling Solution Type Junction Prix Quantity & Availability  
13620151
Accessoires Afficher les documents Détails de la promotion
Thermo Scientific™
GD9106BNWP
Gel-filled Double
chaque for $805.72
Il en reste null
 
13620153
Accessoires Afficher les documents Détails de la promotion
Thermo Scientific™
GD9156BNWP
Liquid-filled Double
chaque for $1,004.39
Il en reste null
 
13620150
Accessoires Afficher les documents Détails de la promotion
Thermo Scientific™
GS9106BNWP
Gel-filled Single
chaque for $636.73
Il en reste null
 
Description

Description

Thermo Scientific™ Orion™ Green Electrodes are the first lab electrode to contain no lead, mercury or other hazardous substances and are completely ROHS compliant. GD9156BNWP is a refillable electrode that provides reliable and accurate measurements of routine samples but are best suited for treated effluent, distilled water and wastewater. GD9106BNWP and GS9106BNWP contain a low-maintenance gel that is best suited for field or plant use and educational institutes, where minimal maintenance is required.
 

Orion™ Double Junction Electrodes have an isolated Ag/AgCl reference system which prevents silver from coming in contact with the sample. Great for measuring TRIS buffer, sulfide and protein samples. While Orion™ Single Junction Electrodes provide a reliable and accurate option for routine pH measurement of typical samples.
 
Solutions required 
Electrode fill solution for GD9156BNWP
910008     3M KCl filling solution, 5 x 60 mL bottles 
 
Electrode storage solution
910001      pH electrode storage solution, 475 mL bottle
 
pH buffers
910168     pH 1.68 buffer, NIST traceable, 475 mL
910104     pH 4.01 buffer, color coded red, NIST traceable, 475 mL
910105     pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
910686     pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
910107     pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL 
910918     pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
910110     pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
910112     pH 12.46 buffer, NIST traceable, 475 mL
910199     Standard All-in-One pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475ml each, pH
                 electrode storage solution, 475ml, and 12 mm electrode storage bottle
 
pH cleaning solution kit
900020     Cleaning solution A and C, 30 ml each. B and D, 60 ml each
 
Items included with GD9156BNWP
910008     3M KCl filling solution 60mL 1 bottle
 
Warranty period
GD9156BNWP: 12 months from date of purchase
GD9106BNWP and GS9106BNWP: 6 months from date of purchase

Warranty

  • 12 months from date of purchase (GD9156BNWP)
  • 6 months from date of purchase (GS9106BNWP and GD9106BNWP).

Spécifications

Spécifications

Epoxy Body
0°C to 90°C
39.3 in.
0 to 14
Orion pH Meter
1 m
Videos
FDS
Documents

Documents