missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ Combination pH Electrodes

Catalog No. 13620151
Change view
Click to view available options
Filling Solution Type:
Gel-filled
Liquid-filled
Junction:
Double
Single
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Filling Solution Type Junction
13620151 Gel-filled Double
13620153 Liquid-filled Double
13620150 Gel-filled Single
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 13620151 Supplier Thermo Scientific™ Supplier No. GD9106BNWP
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Environmentally friendly lab pH electrodes to meet RoHS requirements by containing no lead, mercury or other hazardous substances — making disposal of old electrodes easy and hassle free.

Thermo Scientific™ Orion™ Green Electrodes are the first lab electrode to contain no lead, mercury or other hazardous substances and are completely ROHS compliant. GD9156BNWP is a refillable electrode that provides reliable and accurate measurements of routine samples but are best suited for treated effluent, distilled water and wastewater. GD9106BNWP and GS9106BNWP contain a low-maintenance gel that is best suited for field or plant use and educational institutes, where minimal maintenance is required.
 

Orion™ Double Junction Electrodes have an isolated Ag/AgCl reference system which prevents silver from coming in contact with the sample. Great for measuring TRIS buffer, sulfide and protein samples. While Orion™ Single Junction Electrodes provide a reliable and accurate option for routine pH measurement of typical samples.

Solutions required:

Electrode fill solution for GD9156BNWP

  • 910008 3M KCl filling solution, 5 x 60 mL bottles

Electrode storage solution

  • 910001 pH electrode storage solution, 475 mL bottle

pH buffers

  • 910168 pH 1.68 buffer, NIST traceable, 475 mL
  • 910104 pH 4.01 buffer, color coded red, NIST traceable, 475 mL
  • 910105 pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
  • 910686 pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
  • 910107 pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL
  • 910918 pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
  • 910110 pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
  • 910112 pH 12.46 buffer, NIST traceable, 475 mL
  • 910199 Standard All-in-One pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475 mL each, pH electrode storage solution, 475ml, and 12 mm electrode storage bottle

pH cleaning solution kit

  • 900020 Cleaning solution A and C, 30 mL each. B and D, 60 mL each

Items included with GD9156BNWP

  • 910008 3M KCl filling solution 60 mL 1 bottle

Warranty period

  • GD9156BNWP: 12 months from date of purchase
  • GD9106BNWP and GS9106BNWP: 6 months from date of purchase

Warranty

  • 12 months from date of purchase (GD9156BNWP)
  • 6 months from date of purchase (GS9106BNWP and GD9106BNWP).

Specifications

Description Low-maintenance gel-filled
Connector Types BNC (Waterproof), BNC-to-USS adapter is required with this product as a onetime purchase. Part Number: 13-641-890
Junction Double
Material Epoxy Body
pH Range 0 to 14
Temperature (Metric) Operating 0°C to 90°C
Type Green Combination pH Electrode, gel-filled
Filling Solution Type Gel-filled
For Use With (Equipment) Standard pH Meter
Length (English) Cable 39.3 in.
Length (Metric) Cable 1 m
Show More Show Less