missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ AquaPro™ Double Junction Combination pH Electrodes

Measure the pH of viscous liquid samples quickly, and accurately, with clog-resistant Orion™ AquaPro™ pH electrodes.

Supplier:  Thermo Scientific™ 9135APWP

 Afficher plus de versions de ce produit

Numéro de catalogue. 13645153


Il en reste null

Description

Description

Thermo Scientific™ Orion™ AquaPro™ Low Maintenance pH Electrodes provide fast response and extended life with the additional advantages of a double junction design. Their clog resistant, open junctions allow for use in difficult samples, while making post-use cleaning, simple. They are available in a variety of different designs, with the standard types being the 9104APWP & the 9156APWP. These have glass and epoxy bodies respectively and are designed for general lab use. The 9135APWP has a uniquely designed, flat sensing bulb that allows for reliable pH measurement on samples such as paper, textiles, skin, meat, cheese, or in agar gel plates. The 9107APMD contains a built-in temperature sensor that accurately measures pH and temperature with just one electrode, so combines the fast response and open junction design of the Orion™ AquaPro™ Electrode with a built-in ATC.
 

Feature improved electrode life, low maintenance, fast response time and clean junctions. Patented reference system and double-junction design prevent silver ions from contacting sample.
  • Measure 0 to 14pH
  • Temperature range: 0° to 60°C (0° to 140°F)
  • Choice of connectors with 1m (3ft.) cable
Spécifications

Identifiants chimiques

Open
0°C to 60°C
±0.02
32°F to 140°F
1 m
0 to 14
Ag/AgCl Double Junction
Always store in pH storage solution, Orion 910001. Either store in a beaker of storage solution or reassemble in the storage bottle the electrode was provided with.
39 in.

Spécifications

Epoxy Body
Open
0 to 14
Combination pH Electrode
Orion pH Meter
±0.02
32°F to 140°F
4.72 in.
120 mm
0.47 in.
BNC (Waterproof)
Epoxy Body
0°C to 60°C
Moist surfaces, solids, semisolids, small sample sizes
Ag/AgCl Double Junction
Always store in pH storage solution, Orion 910001. Either store in a beaker of storage solution or reassemble in the storage bottle the electrode was provided with.
Flat Surface
39 in.
1 m
12 mm
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions