missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ AquaPro™ Double Junction Combination pH Electrodes

Catalog No. p-4529926
Change view
Click to view available options
Connector Types:
BNC (Waterproof)
Mini-DIN (Star)
Material:
Epoxy Body
Glass Body
Type:
Combination pH Electrode
pH/ATC Combination Electrode
Tip Type:
Flat Surface
Standard
Toughened Bulb
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connector Types Material Type Tip Type
13645151 BNC (Waterproof) Glass Body Combination pH Electrode Toughened Bulb
13644966 Mini-DIN (Star) Epoxy Body pH/ATC Combination Electrode Standard
13645153 BNC (Waterproof) Epoxy Body Combination pH Electrode Flat Surface
13645154 BNC (Waterproof) Epoxy Body Combination pH Electrode Standard
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 13645151 Supplier Thermo Scientific™ Supplier No. 9104APWP
Only null left

Measure the pH of viscous liquid samples quickly, and accurately, with clog-resistant Orion™ AquaPro™ pH electrodes.

Thermo Scientific™ Orion™ AquaPro™ Low Maintenance pH Electrodes provide fast response and extended life with the additional advantages of a double junction design. Their clog resistant, open junctions allow for use in difficult samples, while making post-use cleaning, simple. They are available in a variety of different designs, with the standard types being the 9104APWP & the 9156APWP. These have glass and epoxy bodies respectively and are designed for general lab use. The 9135APWP has a uniquely designed, flat sensing bulb that allows for reliable pH measurement on samples such as paper, textiles, skin, meat, cheese, or in agar gel plates. The 9107APMD contains a built-in temperature sensor that accurately measures pH and temperature with just one electrode, so combines the fast response and open junction design of the Orion™ AquaPro™ Electrode with a built-in ATC.
 

Feature improved electrode life, low maintenance, fast response time and clean junctions. Patented reference system and double-junction design prevent silver ions from contacting sample.
  • Measure 0 to 14pH
  • Temperature range: 0° to 60°C (0° to 140°F)
  • Choice of connectors with 1m (3ft.) cable

Specifications

Description Glass Body pH
Connector Types BNC (Waterproof)
Junction Open
Material Glass Body
pH Range 0 to 14
Temperature (Metric) Operating 0°C to 60°C
Type Combination pH Electrode
For Use With (Application) Rugged use
For Use With (Equipment) Orion pH Meter
Internal Reference Ag/AgCl Double Junction
pH Relative Accuracy ±0.02
Storage Requirements Always store in pH storage solution, Orion 910001. Either store in a beaker of storage solution or reassemble in the storage bottle the electrode was provided with.
Temperature (English) Operating 32°F to 140°F
Tip Type Toughened Bulb
Length (English) 4.72 in.
Length (English) Cable 39 in.
Length (Metric) 120 mm
Length (Metric) Cable 1 m
Diameter (English) Body 0.47 in.
Diameter (Metric) Body 12 mm
Show More Show Less