missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ Single Junction Thin Stem Glass Bodied Combination pH Electrode

Top-performance, semi-micro pH electrodes to measure samples as small as 0.2 mL

$994.75

Spécifications

Junction Ceramic
Material Glass Body
pH Range 0 to 14
Temperature (Metric) Operating 0°C to 90°C
Filling Solution Type 4M KCl with Ag/AgCl
voir les spécifications

Produits 1
Numéro de catalogue Numéro du manufacturier. Connector Types Prix Quantity & Availability  
Numéro de catalogue Numéro du manufacturier. Connector Types Prix Quantity & Availability  
13642247
Accessoires Afficher les documents Détails de la promotion
Thermo Scientific™
9103BNWP
BNC (Waterproof)
chaque for $994.75
Il en reste null
 
Description

Description

Thermo Scientific™ Orion™ Silver Chloride Semi-Micro Electrodes provide reliable and accurate measurements of routine samples. They are available in chemically resistant glass bodies and best suited for measuring general samples in test tubes and small containers.
 

Orion™ Single Junction Electrodes provide a reliable option for routine pH measurement. These silver chloride electrodes are available in a variety of styles and provide accurate measurements of typical samples.
 
Solutions required 
Electrode fill solution
900011     4M KCl with Ag/AgCl filling solution, 5 x 60 mL bottles
 
Electrode storage solution
910001     pH electrode storage solution, 475 mL bottle
 
pH buffers
910168     pH 1.68 buffer, NIST traceable, 475 mL
910104     pH 4.01 buffer, color coded red, NIST traceable, 475 mL
910105     pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
910686     pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
910107     pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL 
910918     pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
910110     pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
910112     pH 12.46 buffer, NIST traceable, 475 mL
910199     Standard All-in-One pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475ml each, pH
                 electrode storage solution, 475ml, and 12 mm electrode storage bottle
 
pH cleaning solution kit
900020     Cleaning solution A and C, 30 ml each. B and D, 60 ml each
 
Items included with
900011     4M KCl with Ag/AgCl filling solution 60mL 1 bottle
 
Warranty period
12 months from date of purchase
Spécifications

Spécifications

Ceramic
0 to 14
4M KCl with Ag/AgCl
0.02
16 mm
Glass Body
0°C to 90°C
Ag/AgCl
32°F to 194°F
Videos
FDS
Documents

Documents