missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Omni International Omni Tip™ Plastic Homogenizing Probes

The 12 mm Omni Tip significantly reduces the hassle of disassembly and maintenance associated with traditional stainless-steel probes. Engineered from durable plastic, Omni Tips are economical for single use, minimizing cross-contamination when processing multiple samples.
$1,782.39 - $6,464.45
Specifications
| Product Type | Probe |
|---|---|
| Length (Metric) Probe | 12 mm |
| Autoclavable | Autoclavable |
| Blade Material | Polypropylene |
| Closure Color | Blue |
| Catalog Number | Mfr. No. | Packaging Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Packaging Quantity | Price | Quantity & Availability | |||||
15341137
![]() |
Omni International
OMN301250 |
25/Pk. |
Pack for $1,782.39
|
|
|||||
15341138
![]() |
Omni International
321250 |
50/Pk. |
Pack for $3,405.14
|
|
|||||
15341139
![]() |
Omni International
341250 |
100/Pk. |
Pack for $6,464.45
|
|
|||||
Description
- Suitable for homogenization where sample size or sample hardness dictates that processing with the 7 mm tip is prohibitive.
- Significantly reduces cross-contamination and is virtually maintenance-free.
- Designed for PCR, RNA, and DNA extraction.
- Compatible with Omni TH, THQ, GLH homogenizers, and all automation equipment.
- Omni Tips offer a cost-effective solution for researchers homogenizing samples where contaminants can affect downstream assays.
- Provides a cost-effective solution for researchers homogenizing samples where contaminants may impact downstream assays.
Specifications
| Probe | |
| Autoclavable | |
| Blue |
| 12 mm | |
| Polypropylene |