missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Omni International Omni Tip™ Plastic Homogenizing Probes

Description
- Suitable for homogenization where sample size or sample hardness dictates that processing with the 7 mm tip is prohibitive.
- Significantly reduces cross-contamination and is virtually maintenance-free.
- Designed for PCR, RNA, and DNA extraction.
- Compatible with Omni TH, THQ, GLH homogenizers, and all automation equipment.
- Omni Tips offer a cost-effective solution for researchers homogenizing samples where contaminants can affect downstream assays.
- Provides a cost-effective solution for researchers homogenizing samples where contaminants may impact downstream assays.
Specifications
Specifications
| Product Type | Probe |
| Length (Metric) Probe | 12 mm |
| Autoclavable | Autoclavable |
| Blade Material | Polypropylene |
| Closure Color | Blue |
| Packaging Quantity | 100/Pk. |