missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Office Task Lighting

Numéro de catalogue. 06001048
Change view
Click to view available options
Width (English):
20 in.
38 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Width (English)
06001048 20 in.
06001049 38 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. 06001048 Fournisseur Fisherbrand™ Code fournisseur FSLED.20
Il en reste null

Office task lighting can be added to the bottoms of flipper door cabinets or shelving in the office workstation systems.

  • Magnet Attachment
  • LED bulbs
  • Motion sensing and automatic shutoff features

Spécifications

Style LED Lighting Attachment
Width (English) 20 in.
Width (Metric) 50.8 cm