missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Octanoic Acid, 97%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor O1003200LTBL
| Quantity | 200 L |
|---|---|
| Packaging | Amber Glass Bottle |
Identifiants chimiques
| 124-07-2 | |
| 144.21 g/mol |
Spécifications
| Octanoic Acid | |
| 1 | |
| 239°C | |
| 200 L | |
| 0.97 | |
| Liquid |
| 124-07-2 | |
| Colorless to Light Yellow | |
| Amber Glass Bottle | |
| 144.21 g/mol | |
| Reagent |