missing translation for 'onlineSavingsMsg'
Learn More

Octanoic Acid, 97%, Spectrum™ Chemical

$266.53 - $18,551.26

Chemical Identifiers

CAS 124-07-2
Molecular Weight (g/mol) 144.21 g/mol
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
18612072
View Documents Promotion Details
N/A 100 mL Amber Glass Bottle
Each for $266.53
Only null left
 
18612071
View Documents Promotion Details
N/A 1 L Amber Glass Bottle
Each for $1,009.24
Only null left
 
18612075
View Documents Promotion Details
N/A 4 L Amber Glass Bottle
Each for $1,637.91
Only null left
 
18612073
View Documents Promotion Details
N/A 20 L Amber Glass Bottle
Each for $4,189.09
Only null left
 
18612074
View Documents Promotion Details
N/A 200 L Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

124-07-2
144.21 g/mol

Specifications

124-07-2
Colorless to Light Yellow
144.21 g/mol
Reagent
Octanoic Acid
1
239°C
0.97
Liquid
Videos
SDS
Documents

Documents

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.