missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cell Extraction Buffer
Description
Build Your Own Immunoassay, Cell Analysis, Cell Lysis, Immunoassays, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis
Order Info
Shipping condition: 2-8°C
Spécifications
Spécifications
| Content And Storage | Store at 2-8°C |
| Product Type | Cell Extraction Buffer |
| For Use With (Application) | Western Blot, Sample preparation for ELISA |
| Quantity | 100 mL |
| Shipping Condition | 2-8°C |
For Research Use Only. Not for use in diagnostic procedures.