missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Novagen™ pTriEx™-4 DNA
Description
- Transient vertebrate expression is mediated by CMV immediate early enhancer and promoter
- For expression in insect cells, pTriEx-4 contains flanking baculovirus sequences to permit generation of recombinant baculoviruses using the BacVector™ System
- In baculovirus-infected insect cells, expression is driven by the very late p10 promoter
- Expression in E. coli is regulated by tightly controlled T7lac promoter
- pTriEx recombinant plasmids can be transferred into (DE3) pLacI host that allows IPTG based induction
- Available as an Ek/LIC Cloning Kit
Specifications
Specifications
| Format | Glass bottle |
| Product Type | 4 DNA |
| Content And Storage | Store at -20°C. Ok to freeze. |
| Quantity | 20μg |