missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Novagen™ pTriEx™-1.1 DNA

Catalog No. 708403
Change view
Click to view available options
Quantity:
20μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
708403 20μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 708403 Supplier MilliporeSigma™ Supplier No. 708403

Designed to allow rapid characterization of target genes in multiple expression systems. MilliporeSigma™; Novagen™ pTriEx™-1.1 DNA can be used to test expression in E. coli, insect and vertebrate cells.

  • Transient vertebrate expression is mediated by CMV immediate early enhancer and promoter
  • For expression in insect cells, pTriEx-1.1 contains flanking baculovirus sequences to permit generation of recombinant baculoviruses using the BacVector™ System
  • In baculovirus-infected insect cells, expression is driven by the very late p10 promoter
  • Expression in E. coli is regulated by tightly controlled T7lac promoter
  • pTriEx recombinant plasmids can be transferred into (DE3) pLacI host that allows IPTG based induction

Specifications

Format Glass bottle
Product Type 1.1 DNA
Content And Storage Store at -20°C. Ok to freeze.
Quantity 20μg