missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Novagen™ BetaBlue™ Staining Kit

Catalog No. 710743
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
710743 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 710743 Supplier MilliporeSigma™ Supplier No. 710743

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Provides direct visualization of β-galactosidase in cells or tissues.

EMD Millipore Novagen™ BetaBlue™ Staining Kit enables quick, accurate determination of transfection efficiencies.

Applications:
  • Determination of transfection efficiencies
  • Assessment of stable cell line generation
  • Transgene expression in tissue slices or whole mounts of transgenic animals
The BetaBlue™ Staining Kit provides direct visualization of β-galactosidase reporter expression in isolated cells, tissues, or intact organisms. The kit contains sufficient reagents for 100 staining reactions and includes solutions of the substrate X-Gal (5-bromo-4-chloro-3-indolyl-β-D-galactopyranoside) and Reaction Buffer optimized for rapid, sensitive histochemical staining with minimal background. The exceptional staining seen with the BetaBlue Staining Kit enables quick, accurate determination of transfection efficiencies, assessment of stable cell line generation, and transgene expression in tissue slices or whole mounts of transgenic animals.

Specifications

Content And Storage 2°C to -8°C
Quantity 100 mL