Learn More
Restek™ Non-Polar Deactivated Fused Silica Guard Columns and Transfer Lines
Tested with a comprehensive test mix to ensure high inertness, use with most common solvents
Supplier:  Restek™ RK 10031
Encompass
 Afficher plus de versions de ce produit
Numéro de catalogue. 06708129
Il en reste null
Description
Description
  • Tested with a comprehensive test mix to ensure high inertness
  • Useful for a wide range of applications
  • Use with most common solvents
  • Maximum temperature: 325°C
Spécifications
Identifiants chimiques
GC Guard Column
Non-Polar Deactivation
Spécifications
GC Guard Column
Non-Polar Deactivation
0.02 in.
0.53 mm
196.8 ft.
617°F
Test chromatogram
Useful as Guard Columns, Transfer Lines, or Long Retention Gaps
0.02 in.
0.69 mm
60 m
325°C
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS