missing translation for 'onlineSavingsMsg'
Learn More

Bulwark™ Nomex Flame Resistant Knit Balaclava

Catalog No. 17204221
Catalog No. 17204221 Supplier Bulwark™ Supplier No. HNB4NA RG M

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Nomex Flame Resistant Knit Balaclava. Nomex FR CAT 2 AR/FR balaclava offers protection and comfort in one with soft, protective fabric.

  • Nomex FR CAT 2 FR/AR balaclava offers protection and comfort in one with soft, protective fabric.
  • Certifications Level 1: NFPA 70E
  • Certifications Level 2: Category 2 Protection
  • Hazard: Arc Flash, Flash Fire
  • Primary Closure: Pull On
  • Wash Care: Home Wash
  • Fabric: Flame resistant, 8.4 oz. (285 g/m&superscript_2;) Nomex™ knit
  • Blend: Nomex Aramid
  • Features: Double-layer head single-layer extra-long neck skirt, Soft and comfortable with finished edges throughout, Double Layer: Arc Rating ATPV 15 calories/ cm&superscript_2;, Single Layer: Arc Rating ATPV 6.5 calories/ cm&superscript_2; Protection: ATPV Arc Rating: 6.5
  • Country of Origin: Imported, Made in USA Fabric
  • Gender: Unisex
  • Materials: Nomex Knit
  • Brand: Bulwark™

Specifications

Product Type FR Balaclava
Material Nomex
Color White
Closure Type None
Disposable Reusable