missing translation for 'onlineSavingsMsg'
Learn More

Nitrogen Receiver Solution, Reagents

Numéro de catalogue. 01334690
Change view
Click to view available options
Quantity:
10 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
01334690 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 01334690 Fournisseur Labchem Inc Code fournisseur CN00120010F
Il en reste null

Nitrogen Receiver Solution, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Spécifications

Chemical Name or Material Nitrogen Receiver Solution
Boiling Point 100°C
Color Purple
CAS 10043-35-3, 7732-18-5
Physical Form Liquid
Quantity 10 L
Packaging Cubitainer
Grade Reagent