missing translation for 'onlineSavingsMsg'
Learn More

Nitric Acid, 3% (v/v) in Reagent Alcohol, Ricca Chemical

Catalog No. R5316300500
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Natural Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R5316300500 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R5316300500 Supplier Ricca Chemical Company Supplier No. R5316300500A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 64-17-5 , 67-63-0 , 67-56-1 , 7697-37-2 , 7732-18-5
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Color Colorless
Concentration 3% (v/v)
CAS Min % 2.0, 85.0, 4.0, 4.0
CAS Max % 4.0, 90.0, 6.0, 6.0
Packaging Natural Poly Bottle
Solvent or Matrix Reagent Alcohol
Quantity 500 mL
Physical Form Liquid
Percent Purity 3% V/V
Grade Laboratory
Chemical Name or Material Nitric Acid
Show More Show Less