missing translation for 'onlineSavingsMsg'
Learn More

Dräger™ NIOSH Respirator Cartridges

Catalog No. 296355339
Change view
Click to view available options
For Use With (Equipment):
X-plore™ 3300 and 3500 half mask, x-plore™ 5500 full face mask
Product Type:
Cartridge
Color:
Black/Pink
Filter Type:
P100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Product Type Color Filter Type
296355339 X-plore™ 3300 and 3500 half mask, x-plore™ 5500 full face mask Cartridge Black/Pink P100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 296355339 Supplier Dräger™ Supplier No. 6738034

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Cartridge Type Organic Vapor Cartridge
Certifications/Compliance NIOSH
Color Black/Pink
Filter Type P100
Product Type Cartridge
For Use With (Application) To purify air
For Use With (Equipment) X-plore™ 3300 and 3500 half mask, x-plore™ 5500 full face mask
Quantity 1 Ea.