missing translation for 'onlineSavingsMsg'
Learn More

Niobium pieces, 99.8% (metals basis)

Numéro de catalogue. p-7048206
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AA0024122 100 g
AA0024136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AA0024122 Fournisseur Thermo Scientific Chemicals Code fournisseur 00024122
Il en reste null

CAS: 7440-03-1 | Nb | 92.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 3-1-7440
Molecular Formula Nb
Molecular Weight (g/mol) 92.91
MDL Number MFCD00011126
InChI Key GUCVJGMIXFAOAE-UHFFFAOYSA-N
Synonym columbium, element, niobium-93, powder, foil, rod, vn 1, foil, 0.25mm 0.01in thick, columbio, niobio
PubChem CID 23936
ChEBI CHEBI:33344
IUPAC Name niobium
SMILES [Nb]

Spécifications

Quantity 100 g
Form Pieces
Assay Percent Range 99.8% (metals basis)
Percent Purity 99.8%
Assay (metals basis)
Chemical Name or Material Niobium pieces
EINECSNumber 231-113-5
RTECSNumber QT9900000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only