missing translation for 'onlineSavingsMsg'
Learn More

Nickel, plasma standard solution, Specpure™ Ni 1000μg/mL

CAS: 7440-02-0 | Ni | 58.69 g/mol

$142.41 - $329.04

Chemical Identifiers

CAS 7440-02-0
Molecular Weight (g/mol) 58.69
InChI Key PXHVJJICTQNCMI-UHFFFAOYSA-N
PubChem CID 935
ChEBI CHEBI:28112
IUPAC Name nickel
SMILES [Ni]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA13839AE
View Documents
Thermo Scientific Chemicals
013839AE
100 mL N/A
Only null left
 
AA13839AP
View Documents
Thermo Scientific Chemicals
013839AP
500 mL N/A
Only null left
 
Description

Description

Nickel, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-02-0
PXHVJJICTQNCMI-UHFFFAOYSA-N
CHEBI:28112
[Ni]
58.69
935
nickel

Specifications

7440-02-0 , 7697-37-2 , 7732-18-5
Liquid
MFCD00011137 MFCD06798735
Miscible with water
[Ni]
58.69
935
Nitric acid: 5.0%; Nickel: 0.1%; Water: 94.9%
23 hPa (17mm Hg) at 20°C
Ni
Colorless
PXHVJJICTQNCMI-UHFFFAOYSA-N
nickel
100 mL
CHEBI:28112
Nickel
Videos
Safety and Handling

Safety and Handling

P201-P202-P234-P260-P264b-P271-P281-P285-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c

H290-H314-H335-H350i-H360D

DOTInformation : Hazard Class: 8; Packaging Group: II

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only