Learn More
Nickel AA Standard, Certified, 1000ppm ±10ppm (1mL = 1mg Ni), LabChem™
Supplier: LabChem LC176701
Specifications
| Nickel AA Standard | |
| Poly Bottle | |
| 95.76,3.74,0.5 | |
| 0.99cSt | |
| Liquid | |
| 1000 ppm ±10 ppm (1mL = 1mg Ni) | |
| MFCD00149805 | |
| Green | |
| AOPCKOPZYFFEDA-UHFFFAOYSA-N | |
| nickel(2+) hexahydrate dinitrate | |
| 500 mL | |
| 290.79 |
| 1000ppm (1mL = 1mg Ni) | |
| 13478-00-7 , 7697-37-2 , 13478-00-7 , 7732-18-5 | |
| Passes Test | |
| 1.02g/mL | |
| H12N2NiO12 | |
| Ni(NO3)2·6H2O | |
| UN3264 | |
| Soluble in water | |
| O.O.O.O.O.O.[Ni++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
| 290.79 | |
| 61630 | |
| Certified |
Chemical Identifiers
| 13478-00-7 | |
| 290.79 | |
| AOPCKOPZYFFEDA-UHFFFAOYSA-N | |
| nickel(2+) hexahydrate dinitrate |
| H12N2NiO12 | |
| MFCD00149805 | |
| 61630 | |
| O.O.O.O.O.O.[Ni++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Safety and Handling
GHS H Statement
May be corrosive to metals.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep only in original container.
Do not breathe mist.
Wear protective gloves, protective clothing, eye protection, face protection.
Wear respiratory protection.
Wash exposed skin thoroughly after handling.
Contaminated work clothing should not be allowed out of the workplace.
Absorb spillage to prevent material damage.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Store in corrosive resistant container with a resistant inner liner.
Dispose of contents/container to comply with local, state and federal regulations.
Danger
Recommended Storage : Room Temperature