missing translation for 'onlineSavingsMsg'
Learn More

Nickel AA Standard, Certified, 1000ppm ±10ppm (1mL = 1mg Ni), LabChem™

Catalog No. p-5388672
Change view
Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC176707 125 mL Poly Bottle
LC176701 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC176707 Supplier LabChem Supplier No. LC176707
Only null left

Chemical Identifiers

CAS 13478-00-7 , 7697-37-2 , 13478-00-7 , 7732-18-5
Molecular Formula H12N2NiO12
Molecular Weight (g/mol) 290.79
MDL Number MFCD00149805
InChI Key AOPCKOPZYFFEDA-UHFFFAOYSA-N
PubChem CID 61630
IUPAC Name nickel(2+) hexahydrate dinitrate
SMILES O.O.O.O.O.O.[Ni++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Packaging Poly Bottle
CAS Max % 95.76, 3.74, 0.5
Identification Passes Test
Viscosity 0.99cSt
Density 1.02g/mL
Physical Form Liquid
Concentration 1000 ppm ±10 ppm (1mL = 1mg Ni)
Linear Formula Ni(NO3)2·6H2O
UN Number UN3264
Color Green
Solubility Soluble in water
Quantity 125 mL
Formula Weight 290.79
Grade Certified
Chemical Name or Material Nickel AA Standard, 1000ppm (1mL = 1mg Ni)
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
May be corrosive to metals.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep only in original container.
Do not breathe mist.
Wear protective gloves, protective clothing, eye protection, face protection.
Wear respiratory protection.
Wash exposed skin thoroughly after handling.
Contaminated work clothing should not be allowed out of the workplace.
Absorb spillage to prevent material damage.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Store in corrosive resistant container with a resistant inner liner.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature