Learn More
Thermo Scientific™ NheI (10 U/μL)
The NheI restriction enzyme recognizes G^CTAGC sites and cuts best at 37°C in Tango buffer (Isoschizomers: AsuNHI, BmtI).
Supplier: Thermo Scientific™ ER0972
Description
Conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.
Highlights
- Superior quality – stringent quality control and industry leading manufacturing process
- Convenient color-coded Five Buffer System
- Includes universal Tango buffer for double-digestions
- BSA premixed in reaction buffers
- Wide selection of restriction endonuclease specificities
Applications
- Molecular cloning
- Restriction site mapping
- Genotyping
- Southern Blot
- Restriction fragment length polymorphism (RFLP)
- SNP
Note
Low salt, high glycerol (> 5%) concentrations, pH > 8.0 or a large excess of enzyme may result in star activity. NheI is inhibited by salt concentrations above 100 mM. Supercoiled plasmids may require up to 10-fold more NheI for complete digestion than linear DNAs (e.g. 10 units are required to cleave 1 μg of.)
Identifiants chimiques
| 10 U/μL |
Spécifications
| 10 U/μL | |
| Nhe I | |
| Yes | |
| No | |
| Restriction Enzyme |
| CpG Methylation-Sensitive, Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive | |
| 10X Buffer Tango | |
| 37°C | |
| 2,500 units | |
| Traditional Cloning |
For Research Use Only. Not for use in diagnostic procedures.