missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rat NGF beta ELISA Kit

ELISA

$990.00

Spécifications

Accession Number P25427
Assay Range 20.58 to 15,000 pg/mL
Assay Sensitivity 15 pg/mL
Calibration Range 15 pg/mL to 15000 pg/mL
Assay Solid-phase sandwich ELISA
voir les spécifications

Produits 1
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantity & Availability  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantity & Availability  
ERNGF
Encompass
Afficher les documents Détails de la promotion
Invitrogen™
ERNGF
96 Tests
paquet for $990.00
Il en reste null
 
Description

Description

The Rat Nerve Growth Factor Beta(Rt NGFbeta) ELISA quantitates Rt NGFbeta in rat serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Rt NGFbeta. Principle of the method The Rat NGFbeta solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Nerve growth factor (NGF) influences the survival and differentiation of a variety of neuronal and nonneuronal cells. The actions of this polypeptide result from binding to specific cell surface receptors which are present as both high and low affinity sites. Although both types of receptor bind NGF only the high affinity form is capable of signal transduction. Cross-linking studies with 125I-NGF have shown that the high affinity receptor of rat PC12 cells forms an Mr 158,000 complex with NGF while the low affinity receptor forms an Mr 100,000 complex. Full length cDNA clones for both the human and rat NGF receptor genes have been produced and sequenced, revealing greater than 90% homology between the two species. NGF exhibits nerve growth stimulating activity and is involved in the regulation of growth and the differentiation of sympathetic and certain sensory neurons. Further, NGF stimulates division and differentiation of sympathetic and embryonic sensory neurons. Mutations in the NGF gene have been associated with hereditary sensory and autonomic neuropathy, type 5 (HSAN5), and dysregulation of NGF gene expression is associated with allergic rhinitis.
Spécifications

Spécifications

P25427
15 pg/mL
Solid-phase sandwich ELISA
ELISA
Plasma, Serum, Supernatant
96 assays
Colorimetric Microplate Reader
Ngfb,beta-NGF
<10
HRP
Development
2°C to 8°C
NGF
4 hrs 45 mins
20.58 to 15,000 pg/mL
15 pg/mL to 15000 pg/mL
HRP
Other Proteins
This ELISA kit shows no cross-reactivity with the following cytokines tested: rat CINC-2, CINC-3, CNTF, Fractalkine, IL-1 alpha, IL-1 beta, IL-4, IL-6, IL-10, GM-CSF, IFN-gamma, Leptin, Lix, MCP-1, MIP-3 alpha, TIMP-1, TNF-alpha.
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
310738
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
RUO
Plasma 50 μL, Serum 50 μL, Supernatant 100 μL
Ngfb, beta-NGF
1 hrs 20 mins
Videos
FDS
Documents

Documents