missing translation for 'onlineSavingsMsg'
Learn More
Learn More
New Methylene Blue N, For Reticulocytes, Certified, LabChem™
$78.79 - $253.50
Chemical Identifiers
| CAS | 6586-05-6 |
|---|---|
| InChI Key | OPVUYBOFNBJFGV-UHFFFAOYSA-L |
| PubChem CID | 3083969 |
Chemical Identifiers
| 6586-05-6 | |
| OPVUYBOFNBJFGV-UHFFFAOYSA-L | |
| 3083969 |
Specifications
| Passes Test | |
| 97.9,1.6,0.5 | |
| Soluble in water | |
| OPVUYBOFNBJFGV-UHFFFAOYSA-L | |
| 416.05 | |
| Poly Bottle | |
| Hydrogen chloride; Carbon monoxide; Carbon dioxide; Nitrogen oxides; Sulfur compounds | |
| Blue | |
| Liquid |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage : Room Temperature