missing translation for 'onlineSavingsMsg'
Learn More
Learn More
New Methylene Blue N, For Reticulocytes, Certified, LabChem™
Supplier: LabChem LC176601
Specifications
| Passes Test | |
| For Reticulocytes | |
| 97.9,1.6,0.5 | |
| Soluble in water | |
| OPVUYBOFNBJFGV-UHFFFAOYSA-L | |
| 416.05 | |
| Poly Bottle | |
| Hydrogen chloride; Carbon monoxide; Carbon dioxide; Nitrogen oxides; Sulfur compounds | |
| Blue | |
| Liquid |
| New Methylene Blue N | |
| 6586-05-6 , 6487-48-5 , 6586-05-6 , 1934-16-3 | |
| C18H22ClN3S·0·5ZnCl2 | |
| Passes Test | |
| 3083969 | |
| Certified | |
| 1g/mL | |
| 1g/mL | |
| 500 mL |
Chemical Identifiers
| 6586-05-6 | |
| OPVUYBOFNBJFGV-UHFFFAOYSA-L | |
| 3083969 |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage : Room Temperature