accessibility menu, dialog, popup

UserName

CAS RN 1934-16-3

ClH3CHNSHNCH3CH3NH3C

CAS RN 1934-16-3

IUPAC Name: 3,7-bis(ethylamino)-2,8-dimethyl-5λ⁴-phenothiazin-5-ylium chloride
Synonyms: new methylene blue
Molecular Weight (g/mol): 347.91
Molecular Formula: C18H22ClN3S
InChi Key: NZYCYASKVWSANA-UHFFFAOYSA-M
SMILES: [Cl-].CCNC1=CC2=[S+]C3=CC(NCC)=C(C)C=C3N=C2C=C1C
Color 
  • (1)
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)