missing translation for 'onlineSavingsMsg'
Learn More

New Methylene Blue N, For Reticulocytes, Certified, LabChem™

Supplier:  LabChem LC176607

 View more versions of this product

Catalog No. LC176607


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Passes Test
For Reticulocytes
97.9,1.6,0.5
Soluble in water
OPVUYBOFNBJFGV-UHFFFAOYSA-L
416.05
Poly Bottle
Hydrogen chloride; Carbon monoxide; Carbon dioxide; Nitrogen oxides; Sulfur compounds
Blue
Liquid
New Methylene Blue N
6586-05-6 , 6487-48-5 , 1934-16-3
C18H22ClN3S·0·5ZnCl2
Passes Test
3083969
Certified
1g/mL
1g/mL
125 mL

Chemical Identifiers

6586-05-6
OPVUYBOFNBJFGV-UHFFFAOYSA-L
3083969
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions