missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neutral Red TS, 0.1% (w/v) in 50% Alcohol, Ricca Chemical
$125.80 - $216.43
Chemical Identifiers
| CAS | 553-24-2 , 64-17-5 , 67-56-1 , 7732-18-5 |
|---|---|
| Molecular Formula | C15H17ClN4 |
| Molecular Weight (g/mol) | Mixture |
| InChI Key | PGSADBUBUOPOJS-UHFFFAOYSA-N |
| PubChem CID | 11105 |
| ChEBI | CHEBI:86370 |
Chemical Identifiers
| C15H17ClN4 | |
| PGSADBUBUOPOJS-UHFFFAOYSA-N | |
| CHEBI:86370 |
Specifications
| 553-24-2 , 64-17-5 , 67-56-1 , 7732-18-5 | |
| 57.0, 45.28, 0.11 | |
| C15H17ClN4 | |
| Mixture | |
| CHEBI:86370 | |
| 50% Alcohol | |
| Liquid |
| 54.0, 43.51, 0.11 | |
| 0.1% (w/v) | |
| PGSADBUBUOPOJS-UHFFFAOYSA-N | |
| 11105 | |
| Laboratory | |
| Red | |
| Neutral Red, Test Solution |