missing translation for 'onlineSavingsMsg'
Learn More

Neutral Red TS, 0.1% (w/v) in 50% Alcohol, Ricca Chemical

Catalog No. 52714
Change view
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
52714 120 mL Natural Poly Bottle
527116 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 52714 Supplier Ricca Chemical Company Supplier No. 52714

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 553-24-2 , 64-17-5 , 67-56-1 , 7732-18-5
Molecular Formula C15H17ClN4
Molecular Weight (g/mol) Mixture
InChI Key PGSADBUBUOPOJS-UHFFFAOYSA-N
PubChem CID 11105
ChEBI CHEBI:86370

Specifications

CAS Min % 54.0, 43.51, 0.11
CAS Max % 57.0, 45.28, 0.11
Concentration 0.1% (w/v)
Packaging Natural Poly Bottle
Solvent or Matrix 50% Alcohol
Color Red
Quantity 120 mL
Physical Form Liquid
Grade Laboratory
Chemical Name or Material Neutral Red, Test Solution