missing translation for 'onlineSavingsMsg'
Learn More

Neutral Red TS, 0.1% (w/v) in 50% Alcohol, Ricca Chemical

Supplier:  Ricca Chemical Company 527116

Quantity 500 mL
Packaging Natural Poly Bottle

 View more versions of this product

Catalog No. 527116


Only null left

Description

Description

Specifications

Chemical Identifiers

553-24-2 , 64-17-5 , 67-56-1 , 7732-18-5
Mixture
11105
C15H17ClN4
PGSADBUBUOPOJS-UHFFFAOYSA-N
CHEBI:86370

Specifications

Neutral Red
553-24-2 , 64-17-5 , 67-56-1 , 7732-18-5
57.0, 45.28, 0.11
C15H17ClN4
Mixture
CHEBI:86370
Natural Poly Bottle
Red
Liquid
Test Solution
54.0, 43.51, 0.11
0.1% (w/v)
PGSADBUBUOPOJS-UHFFFAOYSA-N
11105
Laboratory
50% Alcohol
500 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions