missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Naphthylethylenediamine Dihydrochloride, 0.1% (w/v), Ricca Chemical
APHA for Nitrite Nitrogen Testing
$111.63 - $195.30
Chemical Identifiers
| Molecular Formula | C12H16Cl2N2 |
|---|---|
| InChI Key | MZNYWPRCVDMOJG-UHFFFAOYSA-N |
Chemical Identifiers
| C12H16Cl2N2 | |
| MZNYWPRCVDMOJG-UHFFFAOYSA-N |
Specifications
| Brown to Green-Yellow | |
| 98.0,0.1 | |
| C12H16Cl2N2 | |
| MZNYWPRCVDMOJG-UHFFFAOYSA-N | |
| N'-naphthalen-1-ylethane-1,2-diamine;dihydrochloride | |
| 15106 | |
| Laboratory | |
| Aqueous Solution | |
| Liquid | |
| Naphthylethylenediamine Dihydrochloride |
| 1465-25-4 , 7732-18-5 | |
| 100.0,0.1 | |
| n-1-naphthyl ethylenediamine dihydrochloride, n1-naphthalen-1-yl ethane-1,2-diamine dihydrochloride, marshall's reagent, n-1-naphthylethylenediamine dihydrochloride, ccris 425, unii-h734599kjl, 1,2-ethanediamine, n-1-naphthalenyl-, dihydrochloride, 2-1-naphthylamino ethylamine 2hcl, bratton-marshall reagent, n-1-naphthalenyl-1,2-ethanediamine dihydrochloride | |
| C1=CC=C2C(=C1)C=CC=C2NCCN.Cl.Cl | |
| 259.174 | |
| CHEBI:53452 | |
| Amber Glass Bottle | |
| 120 mL | |
| 0.1% (w/v) |