missing translation for 'onlineSavingsMsg'
Learn More

Promega Nano-Glo Live Cell Assay System

Numéro de catalogue. PRN2013
Encompass
Change view
Click to view available options
Quantity:
10,000 Assays
100 Assays
1000 Assays
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
PRN2013 10,000 Assays
PRN2011 100 Assays
PRN2012 1000 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. PRN2013 Fournisseur Promega Code fournisseur N2013

Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

The Nano-Glo™ Live Cell Assay System is a single-addition, non-lytic detection reagent used to measure NanoBiT or NanoLuc luminescence from living cells.

  • Measures NanoLuc(R) Luminescence in Living Cells
  • Substrate and buffer optimized to enhance stability
  • Use to detect NanoBiT(R) protein complementation or NanoLuc(R) reporter activity
  • Monitors luminescence at a single time point or continuously for up to 2 hours without compromising cell viability.

Spécifications

Content And Storage -30°C to -10°C
For Use With (Application) NanoBiT™ protein complementation or NanoLuc™ reporter activity
Includes Substrate and Buffer
Quantity 10,000 Assays