Learn More
Thermo Scientific™ Nalgene™ Wide-Mouth PMP Bottles with Closure
GREENER_CHOICE
Carry out a wide array of applications with chemically-resistant Thermo ScientificTM NalgeneTM Wide-Mouth PMP Bottles.
$354.00 - $567.21
Specifications
Color Clear
Description Wide-Mouth Bottle
Material PMP
No. per Pack 4
Sterility Non-sterile
View More Specs
Products 3
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Quantity & Availability  
0292420C
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
21070008
250 mL 43 mm
Pack of 4 Each for $354.00
Case of 6 Pack for $2,124.00
Only null left
0292420E
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
21070016
500 mL 48 mm
Pack of 4 Each for $422.34
Case of 4 Pack for $1,689.36
Only null left
0292420G
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
21070032
1 L 53 mm
Pack of 4 Each for $567.21
Case of 4 Pack for $2,268.84
Only null left
Description
Description
Carry out a wide array of applications with chemically-resistant Thermo Scientific™ Nalgene™ Wide-Mouth PMP Bottles that easily withstand a temperature range from 20° to 175°C (68° to 347°F) and repeated autoclaving even at 150°C (302°F). Made from high quality laboratory grade plastic materials for reliably low leachables and extractables.
  • Designed to tolerate a wide range of temperatures
  • Resistant to acids and bases
  • Better clarity than other bottles of similar chemical resistance
  • Linerless caps for leakproof† performance
Recycling Information: Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged.
Specifications
Specifications
Clear
PMP
Non-sterile
Yes
Round
Unlined
Screw Cap
Autoclavable
Wide-Mouth Bottle
4
Natural
Wide
Wide Mouth Bottle
Polypropylene
No
Videos
SDS
Documents
Documents
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.