missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ PETG Biotainer™ Bottles
GREENER_CHOICE

Numéro de catalogue. 02540261
Change view
Click to view available options
Capacity (Metric):
125 mL
500 mL
1 L
5 L
Product Type:
Square Bottle
Quantity:
14 x 5 Bottles
20 x 5 Bottles
35 Bottles
6 Bottles
7 x 5 Bottles
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Capacity (Metric) Product Type Quantity
02540261 500 mL Square Bottle 14 x 5 Bottles
02540260 125 mL Square Bottle 20 x 5 Bottles
02540263 1 L Square Bottle 7 x 5 Bottles
02540264 1 L Square Bottle 35 Bottles
02540701 5 L Square Bottle 6 Bottles
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Numéro de catalogue. 02540261 Fournisseur Thermo Scientific™ Code fournisseur 300542
Il en reste null

Eliminate the cost of cleaning, sterilizing and associated validations compared to bottles designed for reuse with Thermo Scientific™ Nalgene™ PETG Biotainer™ Bottles, ideal for prolonged storage of pH sensitive materials such as culture media.

Eliminate the cost of cleaning, sterilizing and associated validations compared to bottles designed for reuse with Thermo Scientific™ Nalgene™ PETG Biotainer™ Bottles, ideal for prolonged storage of pH sensitive materials such as culture media. Certified sterile and ready-to-use, all Biotainer bottles are gamma irradiated to a Sterility Assurance Level (SAL) of 10–6.

  • Durable, crystal clear PETG for sample integrity
  • Closures with polyethylene foam core liners for leakproof† assurance
  • Custom options available for fluid transfer applications
  • Supported by extensive validation documentation
  • Suitable for storage in conditions from -40° to +70°C

Warranty: 90 days

Spécifications

Capacity (Metric) 500 mL
Capacity (English) 16.9 oz.
Color Clear
Diameter (English) 3 in.
Diameter (English) Inner Neck 1.1 in.
Diameter (Metric) Inner Neck 28 mm
For Use With (Application) Media/Buffer Preparation ​, Media/Buffer Storage, Sampling, Storage, Transportation ​
Material PET
No. per Pack 5/Pk.
Quantity 14 x 5 Bottles
Sterility Sterile
Temperature Range (Metric) -70°C to 70°C
Certifications/Compliance USP Class VI, Non-cytotoxic, USP <661>, Non-pyrogenic, FDA 21 CFR 177.1315
Closure Included Yes
Graduated Yes
Mouth Narrow
Shape Square
Height (Metric) 176 mm
Product Type Square Bottle
Height (English) 6.93 in.
Closure Size 38 mm
Diameter (Metric) 77 mm
Autoclavable Not Autoclavable
Workflow Step Upstream process solution storage. Upstream Validation
Afficher plus de résultats Afficher moins de résultats

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.