Learn More
Thermo Scientific™ Nalgene™ Opaque Amber HDPE Diagnostic Bottles with Closure: Bulk Pack

Safely store and ship light-sensitive, small-volume valuable reagents with these opaque amber HDPE diagnostic bottles, for use in test kits and many small-volume applications.
Supplier: Thermo Scientific™ 3120049025
Description
Safely store and ship light-sensitive, small-volume valuable reagents with Thermo Scientific™ Nalgene™ Opaque Amber HDPE Diagnostic Bottles. For use in test kits and many small-volume applications, these high density polyethylene bottles include linerless opaque amber polypropylene closures that ensures performance without wrinkles that can cause leaks or contamination of your solutions. The durable, nonpyrogenic and noncytotoxic uniform walls are resistant to splitting or puncturing.
- Nonsterile
Ordering Information:
Bottles and closures are packed in separate polybags within the same carton. Packed 2,000 per case.
Identifiants chimiques
| Opaque Amber | |
| 2000 | |
| Non-sterile | |
| Yes | |
| Sample Container |
| HDPE | |
| 2000 | |
| Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. Product meets the requirements for light-resistant containers per USP latest edition. | |
| Narrow |
Spécifications
| 0.25 oz., 8 mL | |
| 0.25 oz. | |
| 14 mm | |
| 2000 | |
| Non-sterile | |
| Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. Product meets the requirements for light-resistant containers per USP latest edition. | |
| No | |
| Round | |
| Sample Container | |
| Unlined | |
| 20-415 mm | |
| Screw Cap |
| 8 mL | |
| Opaque Amber | |
| HDPE | |
| 2000 | |
| 20-415 mm | |
| Yes | |
| Narrow | |
| 45 mm | |
| 1.58 in. (With Closure), 1.52 in. (Without Closure) | |
| Polypropylene | |
| 25 mm |
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.