Learn More
Thermo Scientific™ Nalgene™ Carboy Replacement Screw Closures and Gaskets
GREENER_CHOICE
Secure-fitting Thermo Scientific™ Nalgene™ Replacement Screw Closures and Gaskets for Nalgene Carboys provide leakproof† service
$124.94 - $891.04
Spécifications
Color White
voir les spécifications
Produits 10
Numéro de catalogue Numéro du manufacturier. Neck Finish Material Product Type Prix Quantity & Availability  
Numéro de catalogue Numéro du manufacturier. Neck Finish Material Product Type Prix Quantity & Availability  
0292314HH
GREENER_CHOICE
Afficher les documents Détails de la promotion
Thermo Scientific™
7121501000
100-400 Polypropylene Screw Cap
caisse de 12 chaque for $330.66
Il en reste null
0292314KK
GREENER_CHOICE
Afficher les documents Détails de la promotion
Thermo Scientific™
7121551200
120-400 Polypropylene Screw Cap
caisse de 12 chaque for $509.36
Il en reste null
02945122
GREENER_CHOICE
Afficher les documents Détails de la promotion
Thermo Scientific™
7122401053
53B HDPE Screw Cap with Strap
caisse de 10 chaque for $891.04
Il en reste null
0292314N
GREENER_CHOICE
Afficher les documents Détails de la promotion
Thermo Scientific™
7121600530
53B Polypropylene Screw Cap
caisse de 12 chaque for $712.68
Il en reste null
12565978
GREENER_CHOICE
Afficher les documents Détails de la promotion
Thermo Scientific™
7121510053
53B HDPE Screw Closure and Gasket
caisse de 12 chaque for $683.29
Il en reste null
012913
GREENER_CHOICE
Afficher les documents Détails de la promotion
Thermo Scientific™
7121600053
53B TPE Sealing Gasket
caisse de 12 chaque for $124.94
Il en reste null
0292314T
GREENER_CHOICE
Afficher les documents Détails de la promotion
Thermo Scientific™
7121600830
83B Polypropylene Screw Cap
caisse de 2 chaque for $361.50
Il en reste null
09548252
GREENER_CHOICE
Afficher les documents Détails de la promotion
Thermo Scientific™
7121621830
83B TPE Sealing Gasket
caisse de 5 chaque for $205.14
Il en reste null
12565979
GREENER_CHOICE
Afficher les documents Détails de la promotion
Thermo Scientific™
7121510083
83B HDPE Screw Cap
caisse de 2 chaque for $343.71
Il en reste null
14511220A
GREENER_CHOICE
Afficher les documents Détails de la promotion
Thermo Scientific™
7122271020
83B Silicone Screw Cap
caisse for $180.04
Il en reste null
Description
Description
Secure-fitting Thermo Scientific™ Nalgene™ Replacement Screw Closures and Gaskets for Nalgene Carboys provide leakproof† service
  • Made to fit Nalgene Carboys
  • Leakproof† when used with the Nalgene bottles with the matching finish
  • Spécifications
    Spécifications
    White
    Videos
    FDS
    Documents
    Documents
    † The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.