missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Carboy Replacement Screw Closures and Gaskets
GREENER_CHOICE

Catalog No. 0292314HH
Change view
Click to view available options
Neck Finish:
100-400
120-400
53B
83B
Material:
HDPE
Polypropylene
Silicone
TPE
Product Type:
Screw Cap
Screw Cap with Strap
Screw Closure and Gasket
Sealing Gasket
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish Material Product Type
0292314HH 100-400 Polypropylene Screw Cap
0292314KK 120-400 Polypropylene Screw Cap
02945122 53B HDPE Screw Cap with Strap
0292314N 53B Polypropylene Screw Cap
12565978 53B HDPE Screw Closure and Gasket
012913 53B TPE Sealing Gasket
0292314T 83B Polypropylene Screw Cap
09548252 83B TPE Sealing Gasket
12565979 83B HDPE Screw Cap
14511220A 83B Silicone Screw Cap
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 0292314HH Supplier Thermo Scientific™ Supplier No. 7121501000
Only null left

Secure-fitting Thermo Scientific™ Nalgene™ Replacement Screw Closures and Gaskets for Nalgene Carboys provide leakproof† service

Secure-fitting Thermo Scientific™ Nalgene™ Replacement Screw Closures and Gaskets for Nalgene Carboys provide leakproof† service

  • Made to fit Nalgene Carboys

Specifications

Color White
Description Replacement Closure
Material Polypropylene
No. per Case 12
Neck Finish 100-400
Mouth Narrow, Wide
Shape Round
Product Type Screw Cap
For Use With (Equipment) Nalgene wider-mouth bottles

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.