missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Fmoc-L-aspartic acid 4-allyl ester, 98%
CAS: 146982-24-3 | C22H21NO6 | 395.411 g/mol
$58.67 - $645.89
Identifiants chimiques
| CAS | 146982-24-3 |
|---|---|
| Molecular Formula | C22H21NO6 |
| Molecular Weight (g/mol) | 395.411 |
| MDL Number | MFCD00190874 |
| InChI Key | FBNFRRNBFASDKS-IBGZPJMESA-N |
| Synonym | fmoc-asp oall-oh, fmoc-l-aspartic acid 4-allyl ester, fmoc-asp oallyl-oh, fmoc-asp oall oh, fmoc-asp-allyl ester, ambotzfaa1353, fmoc-asp oal-oh, pubchem12449, n-fmoc-l-aspartic acid 4-allyl ester, fmoc-asp oall-oh hplc |
| PubChem CID | 10715692 |
| IUPAC Name | (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-oxo-4-prop-2-enoxybutanoic acid |
| SMILES | C=CCOC(=O)CC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13 |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantity & Availability | |||||
|
AAH6358503
|
N/A | 1 g |
chaque for $58.67
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
|
AAH6358506
|
N/A | 5 g |
chaque for $165.90
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
|
AAH6358514
|
N/A | 25 g |
chaque for $645.89
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Slightly soluble in water.
Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Identifiants chimiques
| 146982-24-3 | |
| 395.411 | |
| FBNFRRNBFASDKS-IBGZPJMESA-N | |
| 10715692 | |
| C=CCOC(=O)CC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13 |
| C22H21NO6 | |
| MFCD00190874 | |
| fmoc-asp oall-oh, fmoc-l-aspartic acid 4-allyl ester, fmoc-asp oallyl-oh, fmoc-asp oall oh, fmoc-asp-allyl ester, ambotzfaa1353, fmoc-asp oal-oh, pubchem12449, n-fmoc-l-aspartic acid 4-allyl ester, fmoc-asp oall-oh hplc | |
| (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-oxo-4-prop-2-enoxybutanoic acid |
Spécifications
| 111°C to 115°C | |
| C22H21NO6 | |
| 6670219 | |
| Slightly soluble in water. | |
| C=CCOC(=O)CC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13 | |
| 395.411 | |
| 395.41 | |
| Solid |
| 146982-24-3 | |
| MFCD00190874 | |
| fmoc-asp oall-oh, fmoc-l-aspartic acid 4-allyl ester, fmoc-asp oallyl-oh, fmoc-asp oall oh, fmoc-asp-allyl ester, ambotzfaa1353, fmoc-asp oal-oh, pubchem12449, n-fmoc-l-aspartic acid 4-allyl ester, fmoc-asp oall-oh hplc | |
| FBNFRRNBFASDKS-IBGZPJMESA-N | |
| (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-oxo-4-prop-2-enoxybutanoic acid | |
| 10715692 | |
| 98% | |
| N-Fmoc-L-aspartic acid 4-allyl ester |
Sécurité et manipulation
TSCA : No
missing translation for 'storageNote' : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only