Learn More
N-Fmoc-L-aspartic acid 4-allyl ester, 98%
CAS: 146982-24-3 | C22H21NO6 | 395.411 g/mol
Supplier: Thermo Scientific Chemicals H6358503
| Quantity | 1 g |
|---|
Description
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Slightly soluble in water.
Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Chemical Identifiers
| 146982-24-3 | |
| 395.411 | |
| FBNFRRNBFASDKS-IBGZPJMESA-N | |
| 10715692 | |
| C=CCOC(=O)CC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13 |
| C22H21NO6 | |
| MFCD00190874 | |
| fmoc-asp oall-oh, fmoc-l-aspartic acid 4-allyl ester, fmoc-asp oallyl-oh, fmoc-asp oall oh, fmoc-asp-allyl ester, ambotzfaa1353, fmoc-asp oal-oh, pubchem12449, n-fmoc-l-aspartic acid 4-allyl ester, fmoc-asp oall-oh hplc | |
| (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-oxo-4-prop-2-enoxybutanoic acid |
Specifications
| 111°C to 115°C | |
| 146982-24-3 | |
| MFCD00190874 | |
| fmoc-asp oall-oh, fmoc-l-aspartic acid 4-allyl ester, fmoc-asp oallyl-oh, fmoc-asp oall oh, fmoc-asp-allyl ester, ambotzfaa1353, fmoc-asp oal-oh, pubchem12449, n-fmoc-l-aspartic acid 4-allyl ester, fmoc-asp oall-oh hplc | |
| FBNFRRNBFASDKS-IBGZPJMESA-N | |
| (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-oxo-4-prop-2-enoxybutanoic acid | |
| 395.411 | |
| 395.41 | |
| Solid |
| N-Fmoc-L-aspartic acid 4-allyl ester | |
| C22H21NO6 | |
| 6670219 | |
| Slightly soluble in water. | |
| C=CCOC(=O)CC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13 | |
| 1 g | |
| 10715692 | |
| 98% |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold